accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CNNOCH3CH3SOS

CAS RN 1185885-86-2

IUPAC Name: 2,5-bis(2-ethylhexyl)-3,6-bis(thiophen-2-yl)-1H,2H,4H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione
Synonyms: 2,5-bis(2-ethylhexyl)-3,6-bis(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4-dione
Molecular Weight (g/mol): 524.78
Molecular Formula: C30H40N2O2S2
InChi Key: BTJNHAWTSFHBBN-UHFFFAOYNA-N
SMILES: CCCCC(CC)CN1C(=O)C2=C(N(CC(CC)CCCC)C(=O)C2=C1C1=CC=CS1)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results