accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrBr

CAS RN 118599-27-2

IUPAC Name: 9-benzyl-3,6-dibromo-9H-carbazole
Synonyms: 9-benzyl-3,6-dibromocarbazole
Molecular Weight (g/mol): 415.13
Molecular Formula: C19H13Br2N
InChi Key: QVFWRFBJSIUPAD-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)N(CC1=CC=CC=C1)C1=C2C=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results