accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1186-10-3

IUPAC Name: diethyl (3-bromopropyl)phosphonate
Synonyms: diethyl 3-bromopropylphosphonate
Molecular Weight (g/mol): 259.08
Molecular Formula: C7H16BrO3P
InChi Key: PEIKTSJIUKYDPC-UHFFFAOYSA-N
SMILES: CCOP(=O)(CCCBr)OCC
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results