accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNNHNNONHNNHNNOOOOHHO

CAS RN 1186195-57-2

IUPAC Name: bis(N-cyclohexyl-2-phenoxy-7H-purin-6-amine); oxalic acid
Synonyms: bis(N-cyclohexyl-2-phenoxy-7H-purin-6-amine); oxalic acid
Molecular Weight (g/mol): 708.78
Molecular Formula: C36H40N10O6
InChi Key: ICOKMZZMQHZKGX-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.C1CCC(CC1)NC1=C2NC=NC2=NC(OC2=CC=CC=C2)=N1.C1CCC(CC1)NC1=C2NC=NC2=NC(OC2=CC=CC=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results