CAS RN 118621-30-0
IUPAC Name:
2-(4-bromophenyl)-5-phenylthiophene
Synonyms:
2-(4-bromophenyl)-5-phenylthiophene
Molecular Weight (g/mol):
315.23
Molecular Formula:
C16H11BrS
InChi Key:
YMDCOEOZWHXGTM-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)C1=CC=C(S1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-(4-Bromophenyl)-5-phenylthiophene 98.0+%, TCI America™
CAS: 118621-30-0 Molecular Formula: C16H11BrS Molecular Weight (g/mol): 315.228 MDL Number: MFCD21648452 InChI Key: YMDCOEOZWHXGTM-UHFFFAOYSA-N PubChem CID: 10313583 IUPAC Name: 2-(4-bromophenyl)-5-phenylthiophene SMILES: C1=CC=C(C=C1)C2=CC=C(S2)C3=CC=C(C=C3)Br