accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSNH2OHO

CAS RN 1187-84-4

IUPAC Name: 2-amino-3-(methylsulfanyl)propanoic acid
Synonyms: methylcysteine acm-thiopropionate S-methyl-(9CI)-L-cysteine L-methylcysteine acm-thiopropionic acid
Molecular Weight (g/mol): 135.18
Molecular Formula: C4H9NO2S
InChi Key: IDIDJDIHTAOVLG-UHFFFAOYNA-N
SMILES: CSCC(N)C(O)=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)