accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3SiH3CCH3OOCH3OCH3

CAS RN 118736-04-2

IUPAC Name: tert-butyl[4-(dimethoxymethyl)phenoxy]dimethylsilane
Synonyms: tert-butyl(4-(dimethoxymethyl)phenoxy)dimethylsilane
Molecular Weight (g/mol): 282.46
Molecular Formula: C15H26O3Si
InChi Key: DGWQQAQYHUNLGD-UHFFFAOYSA-N
SMILES: COC(OC)C1=CC=C(O[Si](C)(C)C(C)(C)C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results