accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118754-54-4

IUPAC Name: 2,6-dichloro-4-(trifluoromethoxy)benzaldehyde
Synonyms: 2,6-dichloro-4-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 259.01
Molecular Formula: C8H3Cl2F3O2
InChi Key: OEMRSHNCSCRBQT-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC(Cl)=C(C=O)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results