CAS RN 1187629-43-1
IUPAC Name:
[1,1'-binaphthalene]-2,2'-disulfonyl dichloride
Synonyms:
[1,1'-binaphthalene]-2,2'-disulfonyl dichloride
Molecular Weight (g/mol):
451.33
Molecular Formula:
C20H12Cl2O4S2
InChi Key:
LPHLPJZENXUTJT-UHFFFAOYSA-N
SMILES:
ClS(=O)(=O)C1=CC=C2C=CC=CC2=C1C1=C(C=CC2=CC=CC=C12)S(Cl)(=O)=O