accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2ONHOHO

CAS RN 1188-01-8

IUPAC Name: 2-(2-aminopropanamido)acetic acid
Synonyms: (2-aminopropanamido)acetic acid
Molecular Weight (g/mol): 146.15
Molecular Formula: C5H10N2O3
InChi Key: CXISPYVYMQWFLE-UHFFFAOYNA-N
SMILES: CC(N)C(=O)NCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results