accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1188-02-9

IUPAC Name: (2R)-2-methylheptanoate
Synonyms: (2R)-2-methylheptanoate
Molecular Weight (g/mol): 143.21
Molecular Formula: C8H15O2
InChi Key: NKBWMBRPILTCRD-SSDOTTSWSA-M
SMILES: CCCCC[C@@H](C)C([O-])=O
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)