accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrN

CAS RN 1188365-70-9

IUPAC Name: 6-bromoquinoline-5-carbonitrile
Synonyms: 6-bromoquinoline-5-carbonitrile
Molecular Weight (g/mol): 233.07
Molecular Formula: C10H5BrN2
InChi Key: DTWHKRFZYKUTHB-UHFFFAOYSA-N
SMILES: BrC1=C(C#N)C2=CC=CN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results