accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1189-24-8

IUPAC Name: bis(2-methylpropoxy)(oxo)-λ⁵-phosphanylium
Synonyms: bis(2-methylpropoxy)(oxo)-λ⁵-phosphanylium
Molecular Weight (g/mol): 193.2
Molecular Formula: C8H18O3P
InChi Key: QPKOILOWXGLVJS-UHFFFAOYSA-N
SMILES: CC(C)CO[P+](=O)OCC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results