accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118904-37-3

IUPAC Name: (2R,3S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylpentanoic acid
Synonyms: (2R,3S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-methylpentanoic acid
Molecular Weight (g/mol): 353.42
Molecular Formula: C21H23NO4
InChi Key: QXVFEIPAZSXRGM-KINVLMLONA-N
SMILES: CC[C@H](C)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)