accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1189097-36-6

IUPAC Name: potassium trifluoro(3-iodophenyl)boranuide
Synonyms: potassium trifluoro(3-iodophenyl)boranuide
Molecular Weight (g/mol): 309.91
Molecular Formula: C6H4BF3IK
InChi Key: OREURDWJZUTVFM-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC(I)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results