accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118978-98-6

IUPAC Name: (2-hydroxyethyl)trimethylazanium dimethyl phosphate
Synonyms: choline dimethyl phosphate
Molecular Weight (g/mol): 229.21
Molecular Formula: C7H20NO5P
InChi Key: MCZQEJXSCZNROZ-UHFFFAOYSA-M
SMILES: COP([O-])(=O)OC.C[N+](C)(C)CCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results