accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118994-84-6

NOO

CAS RN 118994-84-6

IUPAC Name: 1,3-oxazole-4-carbaldehyde
Synonyms: 1,3-oxazole-4-carbaldehyde
Molecular Weight (g/mol): 97.07
Molecular Formula: C4H3NO2
InChi Key: JBWIIXBEPINWPB-UHFFFAOYSA-N
SMILES: O=CC1=COC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results