accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118994-89-1

IUPAC Name: ethyl 1,3-oxazole-5-carboxylate
Synonyms: ethyl 1,3-oxazole-5-carboxylate
Molecular Weight (g/mol): 141.13
Molecular Formula: C6H7NO3
InChi Key: KRMORCCAHXFIHF-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results