accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOH

CAS RN 118994-90-4

IUPAC Name: 1,3-oxazole-5-carboxylic acid
Synonyms: 1,3-oxazole-5-carboxylic acid
Molecular Weight (g/mol): 113.07
Molecular Formula: C4H3NO3
InChi Key: QCGMEWVZBGQOFN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results