accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 119-10-8

IUPAC Name: 1-methoxy-4-methyl-2-nitrobenzene
Synonyms: 1-methoxy-4-methyl-2-nitrobenzene
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: LGNMURXRPLMVJI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C)C=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results