accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(R)CH3(R)CH3(R)H3COHH3CO

CAS RN 119-13-1

IUPAC Name: (2R)-2,8-dimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
Synonyms: delta tocopherol delta tocopherol (substance) 8-methyltocol delta-tocopherol preparation delta-tocopherol delta-tocopherol skeleton
Molecular Weight (g/mol): 402.66
Molecular Formula: C27H46O2
InChi Key: GZIFEOYASATJEH-VHFRWLAGSA-N
SMILES: CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=CC(O)=CC(C)=C2O1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results