CAS RN 119-24-4
IUPAC Name:
4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}benzoic acid
Synonyms:
4-{[(2-amino-4-oxo-1H-pteridin-6-yl)methyl]amino}benzoic acid
Molecular Weight (g/mol):
312.29
Molecular Formula:
C14H12N6O3
InChi Key:
JOAQINSXLLMRCV-UHFFFAOYSA-N
SMILES:
NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(O)=O)=CN=C2N1
1
–
2
of
2
results
2
Sigma Aldrich Pteroic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.