accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 119-40-4

IUPAC Name: 4-hydroxy-7-(phenylamino)naphthalene-2-sulfonic acid
Synonyms: 4-hydroxy-7-(phenylamino)naphthalene-2-sulfonic acid
Molecular Weight (g/mol): 315.34
Molecular Formula: C16H13NO4S
InChi Key: YGNDWDUEMICDLW-UHFFFAOYSA-N
SMILES: OC1=C2C=CC(NC3=CC=CC=C3)=CC2=CC(=C1)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results