accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 119-63-1

IUPAC Name: N-(4-aminophenyl)-N-methylacetamide
Synonyms: N-(4-aminophenyl)-N-methylacetamide
Molecular Weight (g/mol): 164.21
Molecular Formula: C9H12N2O
InChi Key: QFELUFGHFLYZEZ-UHFFFAOYSA-N
SMILES: CN(C(C)=O)C1=CC=C(N)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results