accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SOHOO

CAS RN 119-79-9

IUPAC Name: 5-aminonaphthalene-2-sulfonic acid
Synonyms: cleve β-acid cleve acid, mixed mixed cleve's acid kyselina cleve cleve's β-acid 1,6-cleve's acid cleve acid-1,6 1,6-cleve acid cleve's acid-1,6 cleve's acid, mixed
Molecular Weight (g/mol): 223.25
Molecular Formula: C10H9NO3S
InChi Key: UWPJYQYRSWYIGZ-UHFFFAOYSA-N
SMILES: NC1=C2C=CC(=CC2=CC=C1)S(O)(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)