accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 119-84-6

IUPAC Name: 3,4-dihydro-2H-1-benzopyran-2-one
Synonyms: dihydrocoumarin 1,2-benzodihydropyrone dihydrobenzopyrone 3,4-dyhydrocoumarin melilotine hydrocoumarin melilotin melilotol benzodihydropyrone oxochroman
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: VMUXSMXIQBNMGZ-UHFFFAOYSA-N
SMILES: O=C1CCC2=CC=CC=C2O1
Boiling Point 
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)