accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1190-09-6

IUPAC Name: (3-bromopropyl)phosphonate
Synonyms: 3-bromopropylphosphonate
Molecular Weight (g/mol): 200.96
Molecular Formula: C3H6BrO3P
InChi Key: CXOIECRVHUDBSA-UHFFFAOYSA-L
SMILES: [O-]P([O-])(=O)CCCBr
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results