accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1190-48-3

IUPAC Name: (2S)-2-amino-6-formamidohexanoic acid
Synonyms: (2S)-2-amino-6-formamidohexanoic acid
Molecular Weight (g/mol): 174.2
Molecular Formula: C7H14N2O3
InChi Key: KLPJXDPPMSJWKI-LURJTMIESA-N
SMILES: N[C@@H](CCCCNC=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results