accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1190-92-7

IUPAC Name: dimethyl(2-nitroethenyl)amine
Synonyms: dimethyl(2-nitroethenyl)amine
Molecular Weight (g/mol): 116.12
Molecular Formula: C4H8N2O2
InChi Key: JKOVQYWMFZTKMX-UHFFFAOYSA-N
SMILES: CN(C)C=C[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results