accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOO

CAS RN 119001-43-3

IUPAC Name: 4-[bis(4-formylphenyl)amino]benzaldehyde
Synonyms: 4-[bis(4-formylphenyl)amino]benzaldehyde
Molecular Weight (g/mol): 329.36
Molecular Formula: C21H15NO3
InChi Key: YOXHQRNDWBRUOL-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)N(C1=CC=C(C=O)C=C1)C1=CC=C(C=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)