accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNOOHOHO

CAS RN 119062-05-4

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanedioic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanedioic acid
Molecular Weight (g/mol): 355.35
Molecular Formula: C19H17NO6
InChi Key: KSDTXRUIZMTBNV-UHFFFAOYNA-N
SMILES: OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)