accessibility menu, dialog, popup

UserName

Viewing profile as user:
HSSH

CAS RN 1191-08-8

IUPAC Name: butane-1,4-dithiol
Synonyms: 1,4-butanedithiol
Molecular Weight (g/mol): 122.24
Molecular Formula: C4H10S2
InChi Key: SMTOKHQOVJRXLK-UHFFFAOYSA-N
SMILES: SCCCCS
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)