accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1191-50-0

IUPAC Name: sodium tetradecyl sulfate
Synonyms: sodium tetradecyl sulphate
Molecular Weight (g/mol): 316.43
Molecular Formula: C14H29NaO4S
InChi Key: UPUIQOIQVMNQAP-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)