accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 1191-85-1

IUPAC Name: icosa-5,8,11,14-tetraynoic acid
Synonyms: icosa-5,8,11,14-tetraynoic acid
Molecular Weight (g/mol): 296.41
Molecular Formula: C20H24O2
InChi Key: MGLDCXPLYOWQRP-UHFFFAOYSA-N
SMILES: CCCCCC#CCC#CCC#CCC#CCCCC(O)=O
Quantity 
  • (1)
  • (1)
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results