accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1191-95-3

IUPAC Name: cyclobutanone
Synonyms: cyclobutanone
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: SHQSVMDWKBRBGB-UHFFFAOYSA-N
SMILES: O=C1CCC1
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (2)
Quantity 
  • (4)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (5)