accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOClO

CAS RN 1191030-88-2

IUPAC Name: 1-benzofuran-7-sulfonyl chloride
Synonyms: 1-benzofuran-7-sulfonyl chloride
Molecular Weight (g/mol): 216.64
Molecular Formula: C8H5ClO3S
InChi Key: MEOOVUNNRYYGCV-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=C2OC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results