accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOONOO

CAS RN 119137-03-0

IUPAC Name: (2-nitrophenyl)methyl N-cyclohexylcarbamate
Synonyms: (2-nitrophenyl)methyl N-cyclohexylcarbamate
Molecular Weight (g/mol): 278.31
Molecular Formula: C14H18N2O4
InChi Key: NJMCHQONLVUNAM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1COC(=O)NC1CCCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results