accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)(R)H3C

CAS RN 1192-18-3

IUPAC Name: (1R,2S)-1,2-dimethylcyclopentane
Synonyms: (1R,2S)-1,2-dimethylcyclopentane
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: RIRARCHMRDHZAR-KNVOCYPGSA-N
SMILES: C[C@H]1CCC[C@H]1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results