accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1192-37-6

IUPAC Name: methylidenecyclohexane
Synonyms: methylenecyclohexane
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: YULMNMJFAZWLLN-UHFFFAOYSA-N
SMILES: C=C1CCCCC1
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)