accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClON

CAS RN 1192-63-8

IUPAC Name: pyrrolidine-1-carbonyl chloride
Synonyms: pyrrolidine-1-carbonyl chloride
Molecular Weight (g/mol): 133.58
Molecular Formula: C5H8ClNO
InChi Key: XACWJIQLDLUFSR-UHFFFAOYSA-N
SMILES: ClC(=O)N1CCCC1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)