CAS RN 1192045-84-3
IUPAC Name:
2-(2-fluoro-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms:
2-(2-fluoro-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol):
236.09
Molecular Formula:
C13H18BFO2
InChi Key:
XPBIIDZQZBLHMB-UHFFFAOYSA-N
SMILES:
CC1=CC(B2OC(C)(C)C(C)(C)O2)=C(F)C=C1
1
–
1
of
1
results
1
2-Fluoro-5-methylbenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 1192045-84-3 Molecular Formula: C13H18BFO2 Molecular Weight (g/mol): 236.093 InChI Key: XPBIIDZQZBLHMB-UHFFFAOYSA-N Synonym: 2-2-fluoro-5-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-4-fluorobenzene PubChem CID: 56973226 IUPAC Name: 2-(2-fluoro-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane SMILES: B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)C)F