accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1192178-33-8

IUPAC Name: 7a-hydroxy-3a-methyl-octahydro-1H-indene-1,4-dione
Synonyms: 7a-hydroxy-3a-methyl-tetrahydro-2H-indene-1,4-dione
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: CAYWOGVLFCOLCK-UHFFFAOYNA-N
SMILES: CC12CCC(=O)C1(O)CCCC2=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results