accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 119229-99-1

IUPAC Name: 1,1,2,2,3,3,4,4,4-nonafluoro-N-(1,1,2,2,3,3,4,4,4-nonafluorobutanesulfonyl)butane-1-sulfonamide lithium
Synonyms: 1,1,2,2,3,3,4,4,4-nonafluoro-N-(1,1,2,2,3,3,4,4,4-nonafluorobutanesulfonyl)butane-1-sulfonamide lithium
Molecular Weight (g/mol): 588.13
Molecular Formula: C8HF18LiNO4S2
InChi Key: SVSJFMZWEBYTRE-UHFFFAOYSA-N
SMILES: [Li].FC(F)(F)C(F)(F)C(F)(F)C(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results