CAS RN 119237-64-8
IUPAC Name:
(2S)-2-(diphenylmethyl)pyrrolidin-1-ium
Synonyms:
2-diphenylmethylpyrrolidine
Molecular Weight (g/mol):
238.35
Molecular Formula:
C17H20N
InChi Key:
OXOBKZZXZVFOBB-INIZCTEOSA-O
SMILES:
C1C[NH2+][C@@H](C1)C(C1=CC=CC=C1)C1=CC=CC=C1