accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1192541-69-7

IUPAC Name: 4-(6-benzoyl-1H-1,3-benzodiazol-2-yl)benzonitrile
Synonyms: 4-(5-benzoyl-3H-1,3-benzodiazol-2-yl)benzonitrile
Molecular Weight (g/mol): 323.36
Molecular Formula: C21H13N3O
InChi Key: NEPBXFRFQCRWFO-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=C1)C1=CC=C2N=C(NC2=C1)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results