accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CCH3F

CAS RN 1192548-08-5

IUPAC Name: 2-(2-fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(2-fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 236.09
Molecular Formula: C13H18BFO2
InChi Key: WJYURBPLHZZUGJ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(B2OC(C)(C)C(C)(C)O2)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results