CAS RN 1193-12-0
IUPAC Name:
3,3-dimethylpiperidine
Synonyms:
3,3-dimethylpiperidine
Molecular Weight (g/mol):
113.2
Molecular Formula:
C7H15N
InChi Key:
CDODDZJCEADUQQ-UHFFFAOYSA-N
SMILES:
CC1(C)CCCNC1
1
–
1
of
1
results
1
Promotions Available
3,3-Dimethylpiperidine, 95%
3,3-Dimethylpiperidine, 95%
CAS: 1193-12-0 Molecular Formula: C7H15N Molecular Weight (g/mol): 113.20 MDL Number: MFCD00005995 InChI Key: CDODDZJCEADUQQ-UHFFFAOYSA-N PubChem CID: 70942 IUPAC Name: 3,3-dimethylpiperidine SMILES: CC1(C)CCCNC1