accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1193-18-6

IUPAC Name: 3-methylcyclohex-2-en-1-one
Synonyms: 2-cyclohexen-1-one, 3-methyl-
Molecular Weight (g/mol): 110.16
Molecular Formula: C7H10O
InChi Key: IITQJMYAYSNIMI-UHFFFAOYSA-N
SMILES: CC1=CC(=O)CCC1
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (2)
  • (7)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (7)
  • (5)