accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CO

CAS RN 1193-47-1

IUPAC Name: 2,2-dimethylcyclohexan-1-one
Synonyms: cyclohexanone, 2,2-dimethyl-
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: KNSPBSQWRKKAPI-UHFFFAOYSA-N
SMILES: CC1(C)CCCCC1=O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)