accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClON

CAS RN 1193-65-3

IUPAC Name: 1-azabicyclo[2.2.2]octan-3-one hydrochloridyl
Synonyms: 1-azabicyclo[2.2.2]octan-3-one hydrochloridyl
Molecular Weight (g/mol): 160.62
Molecular Formula: C7H11ClNO
InChi Key: IUYBBGKYCRJHHV-UHFFFAOYSA-N
SMILES: [Cl].O=C1CN2CCC1CC2
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
Quantity 
  • (3)
  • (4)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (4)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)